Learn More
Aviva Systems Biology Corp ING1 Antibody - C-terminal region (ARP50512_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50512_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ING1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ING1. Alternate names: p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1aApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60361