Learn More
Aviva Systems Biology Corp ING1 Antibody - N-terminal region (ARP50511_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50511_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ING1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ING1. Alternate names: p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1aApplicationWB. Species reactivity: Human
Catalog No. 50-285-515