Learn More
Aviva Systems Biology Corp ING2 Antibody - N-terminal region (ARP50481_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50481_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ING2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ING2. Alternate names: ING1L, p33ING2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-511