Learn More
Medchemexpress LLC Ingenol 3,20-dibenzoate | 59086-90-7 | 100.0% | 556.65 | 1 ML
SDP
Catalog No. 5000349592
Encompass_Preferred
Ingenol 3,20-dibenzoate is a potent protein kinase C (PKC) isoform-selective agonist. It induces selective translocation of nPKC-delta, -epsilon, and -theta and PKC-mu from the cytosolic to the particulate fraction. It also induces morphologically typical apoptosis through de novo synthesis of macromolecules and increases IFN-γ production and degranulation by NK cells, particularly when NK cells are stimulated by NSCLC cells.
  • Potent protein kinase C (PKC) isoform-selective agonist.
  • Induces selective translocation of nPKC-delta, -epsilon, and -theta and PKC-mu.
  • Induces morphologically typical apoptosis.
  • Increases IFN-γ production and degranulation by NK cells.
  • Lessens anemia induced by 5-fluorouracil in a mouse model.
Catalog No. 50-003-49592 Supplier Medchemexpress LLC Supplier No. HY13729510MM/1ML
Ingenol 3,20-dibenzoate is a potent protein kinase C (PKC) isoform-selective agonist. It induces selective translocation of nPKC-delta, -epsilon, and -theta and PKC-mu from the cytosolic to the particulate fraction. It also induces morphologically typical apoptosis through de novo synthesis of macromolecules and increases IFN-γ production and degranulation by NK cells, particularly when NK cells are stimulated by NSCLC cells.
  • Potent protein kinase C (PKC) isoform-selective agonist.
  • Induces selective translocation of nPKC-delta, -epsilon, and -theta and PKC-mu.
  • Induces morphologically typical apoptosis.
  • Increases IFN-γ production and degranulation by NK cells.
  • Lessens anemia induced by 5-fluorouracil in a mouse model.