Learn More
AdipoGen Ingenol-3-angelate,Chemical. CAS: 75567-37-2
SDP
Supplier:  AdipoGen AGCN20012M001
Encompass_Preferred
Ingenol-3-angelate, AdipoGen Life Sciences. Chemical. CAS: 75567-37-2. Formula: C25H34O6. MW: 430.5. Semi-synthetic. Specific protein kinase C (PKC) activator. Selective activator of PKC isoforms, like PKCtheta in T cells. Antiproliferative and proapoptotic (necrotic). Immunostimulant. Chemotherapeutic. Anticancer compound. Efficacious agent against actinic kerastosis. Antileukemic.
Catalog No. 50-464-315