Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1PLUSCHEM LLC Ingenol mebutate | 75567-37-2 | 430.5 g/mol | 1MG

Catalog No. NC2593421
Encompass_Preferred

Ingenol mebutate | 75567-37-2 | 430.5 g/mol | 1MG

Catalog No. NC2593421 Supplier 1PLUSCHEM LLC Supplier No. 1P00G45E1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ingenol mebutate | 75567-37-2 | 430.5 g/mol | 1MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.