Learn More
AdipoGen Ingenol-3-angelate
SDP
Supplier:  AdipoGen AGCN20012M005
Encompass_Preferred
Chemical. CAS 75567-37-2. Formula C25H34O6. MW 430.5. Semi-synthetic. Specific protein kinase C PKC activator. Selective activator of PKC isoforms, like PKCtheta in T cells. Antiproliferative and proapoptotic necrotic. Immunostimulant. Chemotherapeutic. Anticancer compound. Efficacious agent against actinic kerastosis. Antileukemic.
Catalog No. 50-256-2697