Learn More
Medchemexpress LLC Ingenol-3-palmitate | 52557-26-3 | 99.65% | 586.84 | 1 MG
SDP
Catalog No. 5000351118
Encompass_Preferred
Ingenol-3-palmitate is an ingenane diterpenoid, a natural product isolated from the roots of *Euphorbia ebracteolata*. It is a potent HIV-1 (HIV) inhibitor with an IC50 value of 4.1 nM.
  • Potent HIV-1 (HIV) inhibitor with an IC50 value of 4.1 nM
  • Isolated from the roots of Euphorbia ebracteolata
Catalog No. 50-003-51118 Supplier Medchemexpress LLC Supplier No. HY1372421MG
Ingenol-3-palmitate is an ingenane diterpenoid, a natural product isolated from the roots of *Euphorbia ebracteolata*. It is a potent HIV-1 (HIV) inhibitor with an IC50 value of 4.1 nM.
  • Potent HIV-1 (HIV) inhibitor with an IC50 value of 4.1 nM
  • Isolated from the roots of Euphorbia ebracteolata