Learn More
Medchemexpress LLC Ingenol-3-palmitate | 52557-26-3 | MFCD28252012 | 99.7% | 586.84 | C36H58O6 | 5 MG
SDP
Catalog No. 5000221893
Encompass_Preferred
Ingenol-3-palmitate is an ingenane diterpenoid research compound reported as a potent HIV-1 inhibitor and protein kinase C (PKC) modulator. It is supplied as a colorless to light yellow oil with high purity and is intended for laboratory research use.
  • High purity suitable for research applications.
  • Colorless to light yellow oil physical form.
  • Soluble in DMSO; ultrasonic agitation recommended.
  • Formulatable for in vivo dosing in 10% DMSO/90% corn oil at ≥2.5 mg/mL.
  • Stable under recommended frozen storage conditions.
  • Used in in vitro pharmacology and in vivo formulation studies.
Catalog No. 50-002-21893 Supplier Medchemexpress LLC Supplier No. HY1372425MG
Ingenol-3-palmitate is an ingenane diterpenoid research compound reported as a potent HIV-1 inhibitor and protein kinase C (PKC) modulator. It is supplied as a colorless to light yellow oil with high purity and is intended for laboratory research use.
  • High purity suitable for research applications.
  • Colorless to light yellow oil physical form.
  • Soluble in DMSO; ultrasonic agitation recommended.
  • Formulatable for in vivo dosing in 10% DMSO/90% corn oil at ≥2.5 mg/mL.
  • Stable under recommended frozen storage conditions.
  • Used in in vitro pharmacology and in vivo formulation studies.