Learn More
AdipoGen Ingenol-5,20-acetonide (5 mg)
SDP
Supplier:  AdipoGen AGCN20029M005
Encompass_Preferred
Chemical. CAS: 77573-43-4. Formula: C23H32O5. MW: 388.5. Semisynthetic. Intermediate and starting material for the synthesis and preparation of ingenol 3-ester derivatives. For use in chemical synthesis see references. Has improved shelf life compared to ingenol.
Catalog No. 50-205-3374