Learn More
Medchemexpress LLC HY-N0865 5mg Medchemexpress, Ingenol CAS:30220-46-3 Purity:>98%
SDP
Catalog No. 501933754
Encompass_Preferred
Medchemexpress, HY-N0865 5mg Ingenol CAS:30220-46-3 Ingenol is a PKC activator, with a K i of 30 μM, with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3754 Supplier Medchemexpress LLC Supplier No. HYN08655MG
Medchemexpress, HY-N0865 5mg Ingenol CAS:30220-46-3 Ingenol is a PKC activator, with a K i of 30 μM, with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.