Learn More
Enzo Life Sciences Ingenol (high purity) (5mg). CAS: 30220-46-3
SDP
Supplier:  Enzo Life Sciences ALX350031M005
Encompass_Preferred
Extremely weak protein kinase C (PKC) activator. Starting material for synthesis of ingenol derivatives. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO and 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-8700