Learn More
Medchemexpress LLC HY-B0719 1mg Medchemexpress, Ingenol Mebutate CAS:75567-37-2 Purity:>98%
SDP
Catalog No. 502029404
Encompass_Preferred
Medchemexpress, HY-B0719 1mg Ingenol Mebutate CAS:75567-37-2 Ingenol Mebutate is an active ingredient in Euphorbia peplus, acts as a potent PKC modulator, with Kis of 0.3, 0.105, 0.162, 0.376, and 0.171 nM for PKC-α, PKC-β, PKC-γ, PKC-δ, and PKC-ε, respectively, and has antiinflammatory and antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9404 Supplier Medchemexpress LLC Supplier No. HYB07191MG
Medchemexpress, HY-B0719 1mg Ingenol Mebutate CAS:75567-37-2 Ingenol Mebutate is an active ingredient in Euphorbia peplus, acts as a potent PKC modulator, with Kis of 0.3, 0.105, 0.162, 0.376, and 0.171 nM for PKC-α, PKC-β, PKC-γ, PKC-δ, and PKC-ε, respectively, and has antiinflammatory and antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.