Learn More
Medchemexpress LLC HY-19396 5mg Medchemexpress, Ingliforib CAS:186392-65-4 Purity:>98%
SDP
Catalog No. 501932831
Encompass_Preferred
Medchemexpress, HY-19396 5mg Ingliforib CAS:186392-65-4 Ingliforib is a glycogen phosphorylase inhibitor, with IC 50 s of 52, 352 and 150 nM for liver, muscle and brain glycogen phosphorylase, and has cardioprotective activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2831 Supplier Medchemexpress LLC Supplier No. HY193965MG
Medchemexpress, HY-19396 5mg Ingliforib CAS:186392-65-4 Ingliforib is a glycogen phosphorylase inhibitor, with IC 50 s of 52, 352 and 150 nM for liver, muscle and brain glycogen phosphorylase, and has cardioprotective activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.