Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ IngWAT Mouse Preadipocyte Cell Line

Catalog No. SCC211MI Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Cells:
≥ 1 x 106 Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Cells
SCC211MI ≥ 1 x 106 Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC211MI Supplier MilliporeSigma™ Supplier No. SCC211
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IngWAT mouse immortalized preadipocyte cell line derived from subcutaneous white adipose tissue shown to support induction of adipocytes that express UCP1

  • Derived from subcutaneous white adipose tissue (WAT) isolated from a wild-type mouse
  • Shown to support the induction of beige adipocytes and express UCP1.2 IngWAT mouse immortalized preadipocyte cells are versatile model for a broad spectrum of studies of beige adipocyte induction, maintenance and physiology

Contains genetically modified organisms (GMO)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen, Cell can be cultured at least 10 passages after initial thawing without significantly affecting cell marker expression and functionality
Cell Type Adipocytes
For Use With (Application) Cell Culture
No. of Cells ≥ 1 x 106 Cells/Vial

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.