Learn More
Med Vet International Flurovess (Sevoflurane) Liquid inhalation Anesthetic For Dogs, 250mL, Each
Catalog No. 502828090
Encompass_Preferred
Flurovess (sevoflurane, USP) is indicated for induction and maintenance of general anesthesia in dogs.
Catalog No. 50-282-8090 Supplier Med Vet International Supplier No. RXSEVO250V
Flurovess (sevoflurane, USP) is indicated for induction and maintenance of general anesthesia in dogs.