Learn More
Medchemexpress LLC INH1 | 313553-47-8 | 99.85% | 308.40 | 100 MG
SDP
Catalog No. 5000393484
Encompass_Preferred
INH1 specifically disrupts the Hec1/Nek2 interaction via direct Hec1 binding. It shows promising cancer inhibition activity both in vitro and in vivo. It induces abnormal mitotic processes, as well as cell apoptosis.
  • Specifically disrupts the Hec1/Nek2 interaction via direct Hec1 binding.
  • Exhibits promising cancer inhibition activity both in vitro and in vivo.
  • Induces abnormal mitotic processes and cell apoptosis.
  • Has various IC50 values against different cancer cell lines such as HeLa, K562, MDA-MB-231, and MDA-MB-468.
  • Inhibits tumor outgrowth in a xenografted breast cancer model in nude mice.
  • Available in solid and solution forms.
Catalog No. 50-003-93484 Supplier Medchemexpress LLC Supplier No. HY16660100MG
INH1 specifically disrupts the Hec1/Nek2 interaction via direct Hec1 binding. It shows promising cancer inhibition activity both in vitro and in vivo. It induces abnormal mitotic processes, as well as cell apoptosis.
  • Specifically disrupts the Hec1/Nek2 interaction via direct Hec1 binding.
  • Exhibits promising cancer inhibition activity both in vitro and in vivo.
  • Induces abnormal mitotic processes and cell apoptosis.
  • Has various IC50 values against different cancer cell lines such as HeLa, K562, MDA-MB-231, and MDA-MB-468.
  • Inhibits tumor outgrowth in a xenografted breast cancer model in nude mice.
  • Available in solid and solution forms.