Learn More
Medchemexpress LLC INH14 | 25MG
SDP
Catalog No. 5000307622
Encompass_Preferred
INH14 is a cell permeable inhibitor of IKK /IKK with IC50s of 8 97 and 3 59 M respectively INH14 inhibits the IKK / -dependent TLR inflammatory response INH14 also inhibits downstream of TAK1/TAB1 and NF-kB pathways Anti-inflammatory and anti-cancer activity[1]
Catalog No. 50-003-07622 Supplier Medchemexpress LLC Supplier No. HY11445425MG
INH14 is a cell permeable inhibitor of IKK /IKK with IC50s of 8 97 and 3 59 M respectively INH14 inhibits the IKK / -dependent TLR inflammatory response INH14 also inhibits downstream of TAK1/TAB1 and NF-kB pathways Anti-inflammatory and anti-cancer activity[1]