Learn More
3M™ Inhalation Port Gasket, Replacement Part
Catalog No. 1798596T Shop All 3M Products
Change view
Click to view available options
Quantity:
20/Cs.
Catalog No. Quantity
17-985-96T 20/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 17-985-96T Supplier 3M™ Supplier No. 7000125979
Add to Cart
Add to Cart
This item is not returnable. View return policy
Replacement part for use in side inlet DIN ports of 3M™ Full Facepieces 7800S.
  • Approved 3M replacement part
  • Designed for use in the two side DIN ports of 3M Full Facepieces 7800S (sold separately)
  • Gasket helps prevent leakage through bottom of DIN ports
  • Replacement part can extend service life of facepiece
TRUSTED_SUSTAINABILITY
Specifications
Product Type Replacement Parts and Accessories
For Use With (Application) Facility Maintenance, Cleaning, Assembly and Mechanical, Painting
For Use With (Equipment) 3M™ Full Facepiece 7800 Series
Model 7887
Quantity 20/Cs.