Learn More
Sino Biological Recombinant Human, Mouse, Rat, Cynomolgus, Rhesus, Rabbit Activin A Protein, HPLC-verified
Supplier:  Sino Biological 10429HNAH100
Encompass_Preferred
A DNA sequence encoding mature form of Human / Mouse / Rat / Cynomolgus / Rhesus/ Rabbit INHBA (NP_002183.1) (Gly311-Ser426) was expressed. The mature form sequences of the human, mouse, rat, cynomolgus and rhesus INHBA are identical.
Catalog No. 50-237-1668