Learn More
MyBioSource Human Inhibin beta E (INHBE) ELISA Kit
SDP
Supplier:  MyBioSource MBS202217396STRIPWELLS
Encompass_Preferred
The MyBioSource Human Inhibin beta E (INHBE) ELISA Kit quantifies INHBE in various biological fluids using a 96-well plate coated with a specific antibody. After binding and washing steps, a biotinylated detection antibody and HRP-conjugated streptavidin are added. TMB substrate reveals the level of INHBE by a color change measured at 450 nm.
Catalog No. NC2594258