Learn More
Diagnostic Biosystems INHIBIN Inhibin
Catalog No. 502407467
Encompass_Preferred
Inhibin is a dimeric glycoprotein hormone from the TGF-β family made of α and β subunits. This antibody recognizes the 32kDa α subunit of human inhibin. Inhibin α is expressed in a wide variety of human tissues, such as brain, prostate, adrenal, granulose cells of ovary, sertoli cells of testis and in fetoplacental unit.
Catalog No. 50-240-7467 Supplier Diagnostic Biosystems Supplier No. PDM178
Inhibin is a dimeric glycoprotein hormone from the TGF-β family made of α and β subunits. This antibody recognizes the 32kDa α subunit of human inhibin. Inhibin α is expressed in a wide variety of human tissues, such as brain, prostate, adrenal, granulose cells of ovary, sertoli cells of testis and in fetoplacental unit.