Learn More
Ansh Labs Inhibin A ELISA 96-wells, 50 L Plasma/Serum, Range 6 calibrators ranging 9.9-1188 pg/mL, LOD 5.45 pg/mL, Time 4 hours, Shelf Life 24 months
SDP
Supplier:  Ansh Labs AL123
Encompass
Inhibin A ELISA has a total 4 hours incubation, calibrators and range of 6, 9.9-1188 pg/mL, sensitivity of 5.45 pg/mL, sample of 50 µL Serum. Reacts with Human. Crossreactivity with related molecules is not significant.
Catalog No. 50-194-3731