Learn More
Sino Biological Inhibin beta A / INHBA / Activin A Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 50659R105100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Inhibin beta A / INHBA (rM Inhibin beta A / INHBA; Catalog#50659-M08H; NP_032406.1; Met1-Ser424).
Catalog No. 50-224-7829