Learn More
Charles River Laboratories INHIBITION CARTRIDGE
Catalog No. NC9056669
Encompass
Inhibition/Enhancement Screening Cartridges, 10 single packs of cartridges. Not licensed by the FDA. The disposable cartridge contains four channels - two channels with CSE and LAL, which serve as the positive control channels, and two channels with LAL for testing of samples. To perform the test, add 25 l of sample into all four sample reservoirs.
Catalog No. NC9056669 Supplier Charles River Laboratories Supplier No. PTS220
Inhibition/Enhancement Screening Cartridges, 10 single packs of cartridges. Not licensed by the FDA. The disposable cartridge contains four channels - two channels with CSE and LAL, which serve as the positive control channels, and two channels with LAL for testing of samples. To perform the test, add 25 l of sample into all four sample reservoirs.