Learn More
MSC SKILCRAFT Dust Mop Head, Anti-Microbial, Fits 5" x 24" Frame, Blue
Catalog No. 502839031
Encompass
This dust mop head features antimicrobial yarns to prevent bacteria, mold, and mildew growth, while reducing odors. It uses dust-attracting 100% synthetic fiber looped-end yarn, sewn into pre-shrunk polyester fabric for durability and minimal shrinkage. The yarn contains 25% PET content. Key-hole openings enable easy head changes, and multiple ties secure it to the frame.
  • Antimicrobial yarns prevent bacteria, mold, and mildew growth; reduce odors.
  • Maximum dust-attracting synthetic fiber.
  • Pre-shrunk polyester fabric for multiple washings with minimal shrinkage.
  • Key-hole openings for easy head changes.
  • Multiple ties securely fasten head to frame.
  • Washable.
  • Yarn contains 25% PET content.
  • TAA compliant.
Catalog No. 50-283-9031 Supplier MSC Supplier No. 53408027
This dust mop head features antimicrobial yarns to prevent bacteria, mold, and mildew growth, while reducing odors. It uses dust-attracting 100% synthetic fiber looped-end yarn, sewn into pre-shrunk polyester fabric for durability and minimal shrinkage. The yarn contains 25% PET content. Key-hole openings enable easy head changes, and multiple ties secure it to the frame.
  • Antimicrobial yarns prevent bacteria, mold, and mildew growth; reduce odors.
  • Maximum dust-attracting synthetic fiber.
  • Pre-shrunk polyester fabric for multiple washings with minimal shrinkage.
  • Key-hole openings for easy head changes.
  • Multiple ties securely fasten head to frame.
  • Washable.
  • Yarn contains 25% PET content.
  • TAA compliant.