Learn More
MSC SKILCRAFT Dust Mop Head, Antimicrobial, Looped-End, 5 x 18 in, Red
Catalog No. 502838966
Encompass
The Inhibitor™ Anti-Microbial Dust Mop Head is designed with antimicrobial yarns that effectively prevent the growth of bacteria, mold, and mildew, thereby reducing odors. This mop head is made from maximum dust-attracting 100% synthetic fiber with looped-end yarn, which is sewn into pre-shrunk polyester fabric to ensure minimal shrinkage even after multiple washings. It also incorporates 25% PET content for enhanced durability.
  • Antimicrobial yarns prevent growth of bacteria, mold, and mildew, reducing odors.
  • Maximum dust-attracting 100% synthetic fiber looped-end yarn.
  • Looped-end yarn sewn into pre-shrunk polyester fabric for multiple washings with minimal shrinkage.
  • Yarn contains 25% PET content.
  • Key-hole openings for easy head changes.
  • Multiple ties securely fasten the head to the frame.
Catalog No. 50-283-8966 Supplier MSC Supplier No. 53410460
The Inhibitor™ Anti-Microbial Dust Mop Head is designed with antimicrobial yarns that effectively prevent the growth of bacteria, mold, and mildew, thereby reducing odors. This mop head is made from maximum dust-attracting 100% synthetic fiber with looped-end yarn, which is sewn into pre-shrunk polyester fabric to ensure minimal shrinkage even after multiple washings. It also incorporates 25% PET content for enhanced durability.
  • Antimicrobial yarns prevent growth of bacteria, mold, and mildew, reducing odors.
  • Maximum dust-attracting 100% synthetic fiber looped-end yarn.
  • Looped-end yarn sewn into pre-shrunk polyester fabric for multiple washings with minimal shrinkage.
  • Yarn contains 25% PET content.
  • Key-hole openings for easy head changes.
  • Multiple ties securely fasten the head to the frame.