Learn More
MSC AbilityOne Wet Mop Head, Anti-Microbial, Looped-End, 24 oz, Orange
Catalog No. 502836675
Encompass
These premium mop heads feature new and improved antimicrobial yarns that prevent the growth of bacteria, mold, and mildew while reducing odors. Their looped-end, 4-ply construction ensures superior wear, absorbency, and performance.
  • New and improved antimicrobial yarns with FOSSHIELD, a naturally safe, silver and copper ion infused fiber protected with AgION technology.
  • Prevents the growth of bacteria, mold and mildew while reducing odors.
  • Looped-end, 4-ply construction.
  • Made from rayon, acrylic, and recycled PET polyester fibers.
  • Equipped with a 5-1/2 in nylon mesh headband with six main sews and pre-shrunk tailbands.
  • Washable.
  • Packaged 12 per box.
Catalog No. 50-283-6675 Supplier MSC Supplier No. 53409405
These premium mop heads feature new and improved antimicrobial yarns that prevent the growth of bacteria, mold, and mildew while reducing odors. Their looped-end, 4-ply construction ensures superior wear, absorbency, and performance.
  • New and improved antimicrobial yarns with FOSSHIELD, a naturally safe, silver and copper ion infused fiber protected with AgION technology.
  • Prevents the growth of bacteria, mold and mildew while reducing odors.
  • Looped-end, 4-ply construction.
  • Made from rayon, acrylic, and recycled PET polyester fibers.
  • Equipped with a 5-1/2 in nylon mesh headband with six main sews and pre-shrunk tailbands.
  • Washable.
  • Packaged 12 per box.