Learn More
MSC AbilityOne Anti-Microbial Looped-End Wet Mop Head, 20 oz, Orange
Catalog No. 502838361
Encompass
These premium mop heads feature new and improved antimicrobial yarns infused with silver and copper ions, which prevents the growth of bacteria, mold, and mildew while reducing odors. They have a looped-end, 4-ply construction made from rayon, acrylic, and recycled PET polyester fibers. A 5-1/2 inch nylon mesh headband with six main sews and pre-shrunk tailbands offers superior wear, absorbency, and performance.
  • Antimicrobial protection built directly into the yarn.
  • Crafted from excellent absorbency fibers.
  • Loop-end construction prevents strands from fraying and tangling.
  • Washable.
Catalog No. 50-283-8361 Supplier MSC Supplier No. 53409322
Add to Cart
Add to Cart
These premium mop heads feature new and improved antimicrobial yarns infused with silver and copper ions, which prevents the growth of bacteria, mold, and mildew while reducing odors. They have a looped-end, 4-ply construction made from rayon, acrylic, and recycled PET polyester fibers. A 5-1/2 inch nylon mesh headband with six main sews and pre-shrunk tailbands offers superior wear, absorbency, and performance.
  • Antimicrobial protection built directly into the yarn.
  • Crafted from excellent absorbency fibers.
  • Loop-end construction prevents strands from fraying and tangling.
  • Washable.