Learn More
MSC AbilityOne Wet Mop Head, Anti-Microbial, Looped-End, 24 oz, Red
Catalog No. 502839438
Encompass
These premium mop heads feature new and improved antimicrobial yarns with FOSSHIELD and AgION technology. This advanced design effectively prevents the growth of bacteria, mold, and mildew, reducing odors. The looped-end, 4-ply construction uses rayon, acrylic, and recycled PET polyester fibers, with a nylon mesh headband and pre-shrunk tailbands, ensuring superior wear, absorbency, and performance.
  • Premium anti-microbial design with FOSSHIELD and AgION technology
  • Prevents growth of bacteria, mold, and mildew, reducing odors
  • Superior wear, absorbency, and performance
  • Looped-end, 4-ply construction
  • Made from rayon, acrylic, and recycled PET polyester fibers
  • Nylon mesh headband and pre-shrunk tailbands
  • Washable
Catalog No. 50-283-9438 Supplier MSC Supplier No. 53417192
These premium mop heads feature new and improved antimicrobial yarns with FOSSHIELD and AgION technology. This advanced design effectively prevents the growth of bacteria, mold, and mildew, reducing odors. The looped-end, 4-ply construction uses rayon, acrylic, and recycled PET polyester fibers, with a nylon mesh headband and pre-shrunk tailbands, ensuring superior wear, absorbency, and performance.
  • Premium anti-microbial design with FOSSHIELD and AgION technology
  • Prevents growth of bacteria, mold, and mildew, reducing odors
  • Superior wear, absorbency, and performance
  • Looped-end, 4-ply construction
  • Made from rayon, acrylic, and recycled PET polyester fibers
  • Nylon mesh headband and pre-shrunk tailbands
  • Washable