Learn More
Bachem 4-Aminophenyl-1-thio-b-D-galactopyranoside
SDP
Supplier:  Bachem 4014175.1000
Encompass_Preferred
4-Aminophenyl-1-thio-b-D-galactopyranoside, 29558-05-2, C12H17NO5S, Mr 287.34, 1g. Inhibitor of E.coli b-galactosidase.
Catalog No. 50-194-7040