Learn More
Bachem H-DL-d-Hydroxy-DL-Lys-OH · HCl
SDP
Supplier:  Bachem 4002102.0005
Encompass_Preferred
H-DL-d-Hydroxy-DL-Lys-OH · HCl, 13204-98-3, C6H14N2O3 · HCl, Mr 198.65, 5g. H-DL-Hyl-OH · HCl. Inhibitor of homoserine kinase.
Catalog No. 50-194-7047