Learn More
Bachem H-p-Chloro-DL-Phe-OH
SDP
Supplier:  Bachem 4002988.0025
Encompass_Preferred
H-p-Chloro-DL-Phe-OH, 7424-00-2, C9H10ClNO2, Mr 199.64, 25g. H-4-Chloro-DL-Phe-OH, (RS)-2-Amino-3-(4-chloro-phenyl)-propionic acid, Fenclonine, PCPA. Inhibitor of tryptophan 5-monooxygenase.
Catalog No. 50-194-7080