Learn More
Sigma Aldrich Inhibitor removers
Catalog No. 30632012EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
12 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-632-012EA 12 Ea.
30-632-01EA 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-632-012EA Supplier Sigma Aldrich Supplier No. 30632012EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Inhibitor-remover packings and ready-to-use, disposable prepacked columns conveniently remove small amounts of inhibitors which are added to reagents or solvents that would otherwise be unstable. R: 20-37 S: 22-38-36. Useful in applications which require that the stabilizer or inhibitor (hydroquinone (HQ), hydro-quinone monomethyl ether (MEHQ, 4-methoxyphenol), or 4-tert-butylcatechol (TBC) be removed prior to use. . Columns may be discarded1 when exhausted or reused by filling column approximately 3/4 full with new packing material. HQ/MEHQ columns refill vol.: ∼21g. TBC column refill vol.:∼30g.. Approx. column capacity:HQ/MEHQ: 3L at 100ppmTBC: 4L at 15ppm. TBC columns and packings are not recommended for use with polar solvents, including acrylic acid. Vacuum distillation is suggested for such applications.Easy-to-use:1. Prewashing of prepacked glass columns with a small amount of monomer is recommended.2. Add monomer (or solvent) to an addition funnel which is secured above column.3. Add monomer dropwise to the column. Monitor rate of addition to prevent overflow of the column. Collect monomer in an appropriate container.4. For low-melting solid monomers, keep temperature of column above the melting point during use.Caution: Overheating may cause monomer to polymerize on the column. Viscous or high-melting monomers may be diluted in an appropriate solvent before addition to the column.1Unused columns can be discarded with other solid chemical waste. After use, the toxicity and hazards of the monomer, from which the inhibitor is being removed, should be considered in deciding how to dispose of the packing material or column.Ready-to-use, prepacked, disposable columns for removal of the commonly used inhibitor 4-tert-butylcatechol.
Specifications
Quantity 12 Ea.
MDL Number MFCD00081548