Learn More
Meridian Life Science Inhibitor-Tolerant qPCR Mix, 100ML
SDP
Catalog No. 5000856193
Encompass_Preferred
Meridian’s universal Inhibitor-Tolerant qPCR is a 2x formulation ready-to-use quantitative PCR supermix, containing all components except primers, probes and templates for direct qPCR. It is designed for developing qPCR multiplex assays that require minimal sample processing and fast turn-around times (TAT). Because the mix is highly resistant to various qPCR inhibitors, it can be used with crude lysates or inhibitor-rich samples to directly detect pathogens, including viruses and bacteria, in various biological specimens and to analyze gene expression in cells.
Catalog No. 50-008-56193 Supplier Meridian Life Science Supplier No. MDX013100
Meridian’s universal Inhibitor-Tolerant qPCR is a 2x formulation ready-to-use quantitative PCR supermix, containing all components except primers, probes and templates for direct qPCR. It is designed for developing qPCR multiplex assays that require minimal sample processing and fast turn-around times (TAT). Because the mix is highly resistant to various qPCR inhibitors, it can be used with crude lysates or inhibitor-rich samples to directly detect pathogens, including viruses and bacteria, in various biological specimens and to analyze gene expression in cells.