Learn More
Alkali Scientific Inhibitory Mold Agar 30mL, 15x100mm Plates, 10/Pk
SDP
Catalog No. 5000661943
Encompass_Preferred
Inhibitory Mold Agar (IMA) 30mL Plates, 10/Pk is a selective culture medium formulated for the isolation and recovery of pathogenic fungi from clinical and environmental specimens. The medium supports the growth of molds and yeasts while incorporating antibiotics that suppress bacterial contamination, improving the clarity and reliability of fungal recovery. It is commonly used for detecting medically significant fungi such as Aspergillus, Candida, and Cryptococcus species. The deeper 30mL agar fill provides additional surface depth that supports the development of filamentous fungal colonies during extended incubation. Each plate is prepared under sterile conditions to ensure consistent performance and dependable results in diagnostic, research, and mycology laboratory workflows.
Catalog No. 50-006-61943 Supplier Alkali Scientific Supplier No. IMAP00
Inhibitory Mold Agar (IMA) 30mL Plates, 10/Pk is a selective culture medium formulated for the isolation and recovery of pathogenic fungi from clinical and environmental specimens. The medium supports the growth of molds and yeasts while incorporating antibiotics that suppress bacterial contamination, improving the clarity and reliability of fungal recovery. It is commonly used for detecting medically significant fungi such as Aspergillus, Candida, and Cryptococcus species. The deeper 30mL agar fill provides additional surface depth that supports the development of filamentous fungal colonies during extended incubation. Each plate is prepared under sterile conditions to ensure consistent performance and dependable results in diagnostic, research, and mycology laboratory workflows.