Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Inhibitory Mold Agar
SDP

Catalog No. 23001738 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-738 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-738 Supplier Hardy Diagnostics™ Supplier No. W58
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitory Mold Agar is recommended for the selective isolation of fungi

  • The medium is composed of nutritional factors and inorganic salts which support the growth of most pathogenic fungi
  • Casein and animal tissue provide growth nutrients
  • Yeast extract provides a rich source of vitamins
  • Dextrose, starch and dextrin serve as energy sources
  • Essential minerals and ions are supplied by sodium chloride and metallic salts
  • Gram-positive and gram-negative bacteria are inhibited by chloramphenicol, a broad spectrum antimicrobic

Specifications

Appearance Opalescent
Color Light Amber (small number of fine black flakes may be present)
Content And Storage 2°C to 8°C
Additive Chloramphenicol
Detects Fungi
Form Prepared Medium
Media Type Inhibitory Mold Agar
Product Type Agar Plate
Target Organism Class Fungi
Fill Volume 60 mL
Formulation Yeast Extract - 5.0gm, Dextrose - 5.0gm, Pancreatic Digest of Casein - 3.0gm, Peptic Digest of Animal Tissue - 2.0gm, Starch - 2.0gm, Sodium Phosphate - 2.0gm, Dextrin - 1.0gm, Magnesium Sulfate - 0.8gm, Manganese Sulfate - 0.16gm, Chloramphenicol - 0.125gm, Iron Sulfate - 0.04gm, Sodium Chloride - 0.04gm, Agar - 15.0gm
For Use With (Application) Selective Isolation of Fungi
Incubation Conditions 35°C
Incubation Time Variable
No. per Pack 5
Packaging Type Bag
Quantity 5 Ea.
Dimensions (Dia. x H) 100 x 25 mm
Height (Metric) 25 mm
Depth (Metric) 25 mm
Diameter (Metric) 100 mm
Protein Source Yeast, Casein, Animal Tissue
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.