Learn More
STEMCELL Technologies Cytochalasin D, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 1000556
Encompass
Cytochalasin D is a fungal toxin and a cell-permeable inhibitor of actin polymerization (Brenner & Korn). It also partially blocks HIV-1 release from infected human embryonic stem cells and lymphoma cells (Sasaki et al.).DIFFERENTIATION· Reduces cell viability and inhibits osteoblastic cells differentiation in human stromal stem cells (Chen et al.).CANCER RESEARCH· Binds actin filament and prevents tumor migration in human breast and lung cancer cells (Hayot et al.).
Catalog No. 50-206-9364
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov