Learn More
STEMCELL Technologies Thapsigargin, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 1000569
Encompass
Thapsigargin is a sesquiterpene lactone and a cell-permeable inhibitor of sarco-endoplasmic reticulum Ca2+-ATPases (SERCAs; Sabała et al.; Wictome et al.). It also induces autophagy in mammalian cells (Ding et al.).CANCER RESEARCH· Induces apoptosis in thymocytes in rats (Jiang et al.).· Induces endoplasmic reticulum stress and autophagy in mammalian cells (Ding et al.).
Catalog No. 50-206-9377