Learn More
Med Vet International Lactated Ringer'S Injection, Sterile Nonpyrogenic Solution in Hard Plastic Bottle, 1000mL, Each
Catalog No. 502828812
Encompass_Preferred
This sterile, nonpyrogenic solution is indicated for the correction of electrolyte depletion, metabolic acidosis, and dehydration of cattle, calves, horses, sheep and swine. May be injected intravenously, subcutaneously or intraperitoneally (except in horses) using strict aseptic technique. Each 100 mL contains 600 mg sodium chloride, 310 mg sodium lactate, 30 mg potassium chloride, and 20 mg calcium chloride dihydrate. Packaged in a hard plastic bottle that has a fold-flat integrated hook for hanging.
Catalog No. 50-282-8812 Supplier Med Vet International Supplier No. RXLACTATED1000INJV
This sterile, nonpyrogenic solution is indicated for the correction of electrolyte depletion, metabolic acidosis, and dehydration of cattle, calves, horses, sheep and swine. May be injected intravenously, subcutaneously or intraperitoneally (except in horses) using strict aseptic technique. Each 100 mL contains 600 mg sodium chloride, 310 mg sodium lactate, 30 mg potassium chloride, and 20 mg calcium chloride dihydrate. Packaged in a hard plastic bottle that has a fold-flat integrated hook for hanging.