Learn More
Med Vet International Prevail (Flunixin Meglumine) Injection, 50mg/mL, 250mL, Each
Catalog No. 502828614
Encompass_Preferred
Prevail (flunixin meglumine injection) is indicated in the following:Horse: Prevail is recommended for the alleviation of inflammation and pain associated with musculoskeletal disorders in the horse. It is also recommended for the alleviation of visceral pain associated with colic in the horse.Cattle: Prevail is indicated for the control of pyrexia associated with bovine respiratory disease, endotoxemia and acute bovine mastitis. Prevail is also indicated for the control of inflammation in endotoxemia. See package insert for complete product details.
Catalog No. 50-282-8614 Supplier Med Vet International Supplier No. RXPREVAIL250INJV
Prevail (flunixin meglumine injection) is indicated in the following:Horse: Prevail is recommended for the alleviation of inflammation and pain associated with musculoskeletal disorders in the horse. It is also recommended for the alleviation of visceral pain associated with colic in the horse.Cattle: Prevail is indicated for the control of pyrexia associated with bovine respiratory disease, endotoxemia and acute bovine mastitis. Prevail is also indicated for the control of inflammation in endotoxemia. See package insert for complete product details.