Learn More
Med Vet International Cmpk Sterile Injectable Solution, 500mL, Each
Catalog No. 502828298
Encompass_Preferred
Sterile solution for intravenous use in cattle as an aid in the treatment of conditions associated with calcium, magnesium, phosphorus, potassium or glucose deficiencies. Contains dextrose, calcium borogluconate, potassium chloride, sodium hypophosphite and magnesium borogluconate.
Catalog No. 50-282-8298 Supplier Med Vet International Supplier No. RXCMPK500INJV
Sterile solution for intravenous use in cattle as an aid in the treatment of conditions associated with calcium, magnesium, phosphorus, potassium or glucose deficiencies. Contains dextrose, calcium borogluconate, potassium chloride, sodium hypophosphite and magnesium borogluconate.