Learn More
Med Vet International Contrased (Atipamezole Hydrochloride) Sterile Injectable Solution, 5mg/mL, 10mL, Each
Catalog No. 502828300
Encompass_Preferred
ContraSed Injectable Solution (atipamezole hydrochloride) is indicated for the reversal of the sedative and analgesic effects of dexmedetomidine hydrochloride and medetomidine hydrochloride.
Catalog No. 50-282-8300 Supplier Med Vet International Supplier No. RXCONT510INJV
ContraSed Injectable Solution (atipamezole hydrochloride) is indicated for the reversal of the sedative and analgesic effects of dexmedetomidine hydrochloride and medetomidine hydrochloride.