Learn More
Med Vet International Vitamin C Injectable Solution, 100mL, Each
Catalog No. 502827795
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
For use as a supplement of vitamin C in cattle, horses, sheep, swine, dogs and cats. Administer IM. Each mL contains 250 mg Sodium Ascorbate.
Catalog No. 50-282-7795 Supplier Med Vet International Supplier No. RXVITAC100INJV
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
For use as a supplement of vitamin C in cattle, horses, sheep, swine, dogs and cats. Administer IM. Each mL contains 250 mg Sodium Ascorbate.