Learn More
Med Vet International Vitamin B-12 3000mcg Sterile Solution Injection, 250mL, Each
Catalog No. 502827794
Encompass_Preferred
For use as a supplemental nutritive source of Vitamin B-12 in cattle, horses, sheep, swine, dogs and cats. Inject subcutaneously and/or intramuscularly. Each mL Contains: Cyanocobalamin (Vitamin B-12) 3000mcg Benzyl Alcohol (as preservative) 1.5% v/v Sodium Chloride 0.8% w/v Water for Injection q.s. pH buffered with Acetic Acid
Catalog No. 50-282-7794 Supplier Med Vet International Supplier No. RXVITAB12250INJV
For use as a supplemental nutritive source of Vitamin B-12 in cattle, horses, sheep, swine, dogs and cats. Inject subcutaneously and/or intramuscularly. Each mL Contains: Cyanocobalamin (Vitamin B-12) 3000mcg Benzyl Alcohol (as preservative) 1.5% v/v Sodium Chloride 0.8% w/v Water for Injection q.s. pH buffered with Acetic Acid