Learn More
Kent Scientific Corporation INJECT AND BLOOD COLL NDLS 100
SDP
Supplier:  Kent Scientific Corporation INFHWBI
Encompass
Humaneway Winged Infusion Sets are perfect for researchers seeking precise reliable intravenous access especially when practicing with Humaneway Tail Vein Simulators. The flexible wings allow easy handling and the ultra-sharp needles minimize tissue damage ensuring consistent results.Box includes 100 disposable 25G blood collection needles that can be used by connecting to any size capillary tubes or syringes.
Catalog No. NC3908731