Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTL-Strefa Inject-Safe™ Barrier Bandage
SDP

Catalog No. 22192024
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-192-024 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-192-024 Supplier HTL-Strefa Supplier No. 3070
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Inject-Safe™ Barrier Bandage is a patented pre-injection self-adhesive bandage applied prior to subcutaneous or intramuscular injection, providing containment of post-injection bleeding.

  • Provide your frontline clinicians with an increased level of protection during vaccination administration
    • A patented pre-injection self-adhesive bandage applied prior to subcutaneous or intramuscular injection, providing containment of post-injection bleeding.
  • How DropSafe Inject-Safe Barrier Bandage Protects
    • Innovative protective barrier to limit healthcare provider contact to exposed injection site.
    • Enables healthcare professional to focus on immediate activation of the safety mechanism and safely dispose of the needle and syringe.
    • Eliminates the necessity to return to the injection site.
  • Inject-Safe Barrier bandage has a White outer self-adhering spacer ring that creates a liquid-tight seal against the skin. At the site of injection, the self-sealing, non-coring elastomeric membrane becomes liquid-tight after injection.

Specifications

Material Film, Foam
Sterility Sterile
Length (English) 1.37 in.
Quantity 100 Pk.
For Use With (Application) Containment of Post-Injection Bleeding
Color White
Shape Round
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.