Learn More
Med Vet International Injection Cap, Sterile, Vet Use Only, 24/Pack
Supplier:  Med Vet International INJCAPX24
Encompass_Preferred
Sterile. Single use. Compatible with all I.V. Catheters. Ideal for use as a catheter plug. Luer lock. Vet use only.
Catalog No. 50-209-2301