Learn More
Teledyne ISCO, Inc INJECTION VALVE ASSMBLY,6PORT
Catalog No. NC1888823 Shop All Teledyne ISCO Inc. Products
Encompass
Injection valve assembly, 6 port. Includes valve, flexible coupling, nuts, ferrules and installation instructions.
Catalog No. NC1888823 Supplier Teledyne ISCO, Inc Supplier No. 605234218
Injection valve assembly, 6 port. Includes valve, flexible coupling, nuts, ferrules and installation instructions.