Learn More
Waters Corp Waters Injection Valve Cartridge, H-Class Bio, UPLC System
Catalog No. 502039528 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement injection valve cartridge is used in H-Class Bio Flow through Needle (FTN) Sample Manager and ACQUITY Arc Bio Flow through Needle (FTN) Sample Managers. It is designed for UPLC systems and should be replaced as required, with one unit provided per package.
  • Product type: Spare parts kits
  • System type: UPLC
  • Replacement schedule: As required
  • Units per package: 1 pk
  • Type: Injector/valve pod
  • Compatibility: H-Class Bio Flow through Needle (FTN) Sample Manager and ACQUITY Arc Bio Flow through Needle (FTN) Sample Managers
Catalog No. 50-203-9528 Supplier Waters Corp Supplier No. 700005407
This replacement injection valve cartridge is used in H-Class Bio Flow through Needle (FTN) Sample Manager and ACQUITY Arc Bio Flow through Needle (FTN) Sample Managers. It is designed for UPLC systems and should be replaced as required, with one unit provided per package.
  • Product type: Spare parts kits
  • System type: UPLC
  • Replacement schedule: As required
  • Units per package: 1 pk
  • Type: Injector/valve pod
  • Compatibility: H-Class Bio Flow through Needle (FTN) Sample Manager and ACQUITY Arc Bio Flow through Needle (FTN) Sample Managers